Learn More
BioVendor, LLC ENDOLISA ENDOTOXIN ASSAY (192 TESTS)
SDP
Supplier:  BioVendor, LLC 609033
Encompass
EndoLISA is intended for quantitative determination of endotoxin (chemicallylipopolysaccharide, LPS) in pharmaceutical end-products, in-processcontrol and research samples, and medical device testing. EndoLISAis particularlysuited forcomplex samples. • Overcomes limitations of conventional methods such as inhibition and enhancement• Endotoxin specific, no false-positive resultsfrom ß-Glucans• Highly robust method and excellent lot-to-lotreproducibility• Broad measurement range(0.05-500 EU/ml, pH4-pH9)• Animal-free method relying on sustainableresources• Methodology is included in the EuropeanPharmacopoeia Chapter 5.1.10, Section 9.Removal of Interfering Factors• Validated according to standard BacterialEndotoxin Testing (BET) criteria
Catalog No. NC0546492