Learn More
Apexbio Technology LLC Endomorphin-2 141801-26-5 5mg
SDP
Supplier:  Apexbio Technology LLC B50455
Encompass_Preferred
Endomorphin-2 (CAS 141801-26-5) is a selective agonist of the -opioid receptor (MOR) with high affinity and specificity Studies demonstrate that Endomorphin-2 reduces the frequency and amplitude of spontaneous excitatory postsynaptic currents (sEPSCs) in spinal motoneurons an effect reversed by the MOR antagonist CTOP This modulation involves both presynaptic and postsynaptic mechanisms resulting in altered motor neuron activity Endomorphin-2 is utilized in neurobiological research particularly for investigating mechanisms underlying central nervous system disorders and pain modulation
Catalog No. 50-000-88866
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.