Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Endomucin Monoclonal Antibody (eBioV.7C7 (V.7C7)), eBioscience™, Invitrogen™

Catalog No. 5013211
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-132-11 50 μg
50-132-12 100 μg
50-132-13 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-132-11 Supplier Invitrogen™ Supplier No. 14585181
Only null left
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: The eBioV.7C7 monoclonal antibody reacts with mouse endomucin, which was identified in a search for cell-surface expressed endothelial cell markers. Endomucin is a 75 kDa type I integral membrane protein, with similarities to the sialomucin family of proteins including extensive O-linked glycosylation. Endomucin is expressed on endothelial cells, however, an exception is the high endothelial venules (HEV) of secondary lymphoid organs. In addition, it has been demonstrated that endomucin is expressed on CD34-c-Kit+Sca-1+Lin- hematopoietic progenitors, and that these cells are capable of multi-lineage long-term reconstitution of the hematopoietic compartment. Applications Reported: This eBioV.7C7 (V.7C7) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, immunoblotting (WB), and immunohistology staining of paraffin embedded tissue sections. Applications Tested: This eBioV.7C7 (V.7C7) antibody has been tested by flow cytometric analysis of bEnd.3 cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE.

Endomucin (EMCN) also known as endothelial sialomucin or mucin-like sialoglycoprotein interferes with the assembly of focal adhesion complexes and inhibits interaction between cells and the extracellular matrix.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endomucin
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone eBioV.7C7 (V.7C7)
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Emcn
Gene Accession No. Q9R0H2
Gene Alias 0610012K22Rik; AI315669; Emcn; EMCN2; Endomucin; endomucin-1; endomucin-1/2; Endomucin-2; gastric cancer antigen Ga34; Muc14; MUC-14; mucin 14, endothelial; mucin-14; sialomucin
Gene Symbols Emcn
Host Species Rat
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59308
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.