Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Endonuclease V, T.maritima (5 U/μL)

Catalog No. FEREN0141 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREN0141 250 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEREN0141 Supplier Thermo Scientific™ Supplier No. EN0141
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Degrade DNA with Endonuclease V, T. maritima (Endo V), a 3'-endonuclease involved in DNA repair that initiates removal of deaminated bases from damaged DNA.

Endonuclease V, T. maritima (Endo V) is a 3'-endonuclease involved in DNA repair, which initiates removal of deaminated bases from damaged DNA, including uracil, hypoxanthine, and xanthine (see Figure 1).

Endonuclease V is also active toward abasic sites and urea sites, base pair mismatches, flap and pseudo Y structures, and small insertions/deletions in DNA molecules. The cleavage site generated by Endonuclease V is at the second phosphodiester bond 3' to a lesion. 

Highlights

  • Optimal activity at temperatures of 65 to 70°C
TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Enzyme Endonuclease
Quantity 250 units
Product Type Endonuclease V
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.