Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endonuclease VIII, 2000 U, G-Biosciences
SCI_ED

Catalog No. S001140
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
S001140 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001140 Supplier G-Biosciences Supplier No. 7862274
Add to Cart
Add to Cart

Endonuclease VIII from E. coli has dual functions as an N-glycosylase and an AP-lyase. It removes damaged pyrimidines from double-stranded DNA to create an AP site. The AP-lyase activity then cleaves at this site, resulting in a 5′ phosphate and a 3′ phosphate.

  • This enzyme targets damaged bases like thymine glycol and uracil glycol. Unlike Endonuclease III, which only has β lyase activity, Endonuclease VIII also exhibits δ lyase activity, enabling it to address a broader range of DNA damage.

Specifications

Packaging Vial
Enzyme Endonuclease
Sensitive to Heat Inactivation Yes
Product Type Endonuclease VIII
Research Category DNA Repair
Product Line Thermo Scientific
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.