Learn More
Cygnus Technologies EndonucleaseGTP ELISA Kit
Supplier:  Cygnus Technologies F960
Encompass
Cygnus Technologies EndonucleaseGTP ELISA Kit is a sensitive and specific assay to detect and quantify residual endonuclease impurity in recombinant viral vector and vaccine preparations. Expression of recombinant viral vectors and vaccines in mammalian cell culture is a cost-effective method for production of commercial quantities of gene therapy and vaccine biologics. The manufacturing and purification process of these products leaves the potential for contamination by endonuclease enzyme often used to remove endogenous host cell DNA and RNA, as well as unincorporated plasmid DNA from viral vector production. It is desirable to reduce endonuclease contamination to the lowest levels practical.
Catalog No. NC1743594