Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Endopan 300 SL Kit - Serum-free medium for HUVEC and endothelial cells (1 kit, 500 ml)
SDP
Supplier:  ILEX LIFE SCIENCES LLC P040065K
Encompass_Preferred
PAN-Biotech Endopan 300 SL Kit: Serum-free medium kit for HUVEC and endothelial cells (1 kit, 500 ml). DESCRIPTION: PAN-Biotech Endopan 300 SL is a serum-free medium designed for serum-free culture of endothelial cells directly after isolation; optimized for the maintenance and expansion of endothelial cells under serum-free culture conditions. SUITABILITY: Endopan 300 SL is suitable for the culture of human umbilical vein endothelial cells (HUVEC), which are the cells of choice for studying function and pathology of endothelial cells (e.g. angiogenesis). They can be cultured continuously with a stable growth rate in serum-free media. KIT CONTENTS: Endopan 300 SL Kit contains the following components in single packing: Basal Medium Endopan 300 SL, Serum substitute Panexin SL-S, EGF, FGF-2, VEGF, Vitamin C, R3-IGF-1, GA, Hydrocortisone, and Heparin.
Catalog No. 50-238-8510
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.