Learn More
Genscript Corporation β-Endorphin, human
Supplier:  Genscript Corporation RP11344
Encompass_Preferred
Potent endogenous opioid protein b-Endorphin is derived from propiomelanocortin, b-Endorphin is a protein found in the brain, anterior pituitary, skin, immune system, and other peripheral sites. β-Endorphin is released in response to painful stimuli and b-Endorphin has potent antinociceptive activity mediated through its action on μ receptors in brain and by μ and κ receptors in the spinal cord.
Catalog No. NC1763199
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.