Learn More
Enzo Life Sciences Endothall (100mg). CAS: 145-73-3
SDP
Supplier:  Enzo Life Sciences BMLPR1070100
Encompass_Preferred
Intermediate-potency protein phosphatase 2A (PP2A) inhibitor (IC50=970nM, IC50 for PP1=5000nM). Useful for structure/activity correlations. Alternative name: 7-Oxabicyclo[2.2.1]heptane-2,3-di-carboxylic acid. Purity: ≥98%. Solubility: Soluble in water (>25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1505
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.