Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Endothall

Catalog No. AAJ60948MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60948MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ60948MC Supplier Thermo Scientific Chemicals Supplier No. J60948MC
Get 15% Off Thermo Scientific Reagents
Use promo code REAGENTS15
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

A moderately potent inhibitor of protein phosphatase 2A

Endothall is used as an inhibitor of PP2A phosphatase activity. It is also used as a defoliant. It is used as a cell adhesion inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145-73-3
Molecular Formula C8H10O5
Synonym 7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic acid

Specifications

Quantity 100 mg
Description White
Content And Storage 2°C to 8°C
For Use With (Application) A moderately potent inhibitor of protein phosphatase 2A
Formula Weight 186.20g/mol
Product Type Signal Transduction Reagent
Form Powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.