Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endothall Solution, 50μg/mL in methanol, Ultra Scientific

Catalog No. USPPS2111
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USPPS2111 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USPPS2111 Supplier Agilent Technologies Supplier No. PPS2111
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 145-73-3 , 67-56-1
Molecular Formula C8H8Na2O5
Molecular Weight (g/mol) 230.13
MDL Number MFCD00137362,MFCD00047137
InChI Key XRHVZWWRFMCBAZ-UHFFFAOYNA-L
PubChem CID 3225
IUPAC Name disodium 7-oxabicyclo[2.2.1]heptane-2,3-dicarboxylate
SMILES [Na+].[Na+].[O-]C(=O)C1C2CCC(O2)C1C([O-])=O

Specifications

CAS Max % 99.99, 0.0
Analyte or Component Names Methanol,Endothall
Element Methanol,Endothall
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 50μg/mL
Chemical Name or Material Endothall Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.