Learn More
Description
- Support expansion of human primary endothelial cells under
low-serum conditions - Supports CD31+, KDR+,vWF+, human umbilical vein, endothelial cell growth in vitro
Storage: Store in the dark at 2°C to 8°C. Do not freeze.
Specifications
Specifications
| Product Type | Growth Medium |
| For Use With (Application) | Cell Growth and Differentiation |
| Quantity | 250 mL |
| Content And Storage | Store the unopened product at 2 - 8° C. Do not use past expiration date. |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
