Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Endothelial Cell Growth Media without VEGF
SDP

Catalog No. CCM029 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CCM029 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
When handling biohazardous materials such as human cells, safe laboratory procedures should be followed and protective clothing should be worn. This product is for research use only and is not approved for diagnostic procedures.
Catalog No. CCM029 Supplier R&D Systems™ Supplier No. CCM029

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
When handling biohazardous materials such as human cells, safe laboratory procedures should be followed and protective clothing should be worn. This product is for research use only and is not approved for diagnostic procedures.

Formulated to support the expansion of human primary endothelial cells under low-serum, VEGF-free conditions

  • Unique and optimized formulation that promotes endothelial cell growth.
  • Free of Vascular Endothelial Growth Factor (VEGF)
  • Includes a single supplement featuring key growth factors and small molecules
  • 250 mL bottle results in less wasted media
  • This media has been tested for its ability to support CD31+, KDR+, and vWF+ human umbilical vein endothelial cells growth in vitro
  • This kit contains optimized concentrations of the following reagents to maintain and expand human and mouse endothelial cells in culture:
    • Endothelial Cell Growth Base Media (250 mL)
    • Endothelial Cell Growth Supplement without VEGF (50X)
      • Human FGF basic
      • Human LR3 IGF-1
      • Human EGF
      • Heparin
      • Hydrocortisone
      • L-Ascorbic Acid
      • Fetal Bovine Serum
  • FOR LABORATORY RESEARCH USE ONLY. NOT FOR USE IN DIAGNOSTIC PROCEDURES.

Specifications

With Additives Growth Factors
Product Line R&D Systems
Quantity 250 mL
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.