Learn More
Description
Includes
Human FGF basic, Human LR3 IGF-1, Human VEGF165, Human EGF, Heparin,Hydrocortisone, L-Ascorbic Acid,Fetal Bovine Serum
- Support expansion of human primary endothelial cells under
low-serum conditions - Supports CD31+, KDR+,vWF+, human umbilical vein, endothelial cell growth in vitro
Storage: Store in the dark at 2°C to 8°C. Do not freeze.
Specifications
Specifications
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Product Type | Growth Supplement |
| For Use With (Application) | Cell Growth and Differentiation |
| Product Line | R&D Systems |
| Quantity | 5 mL |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
