Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Endothelin-1 (ET-1) ELISA Kit

Catalog No. EIAET1X10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIAET1X10 10 x 96 Tests
EIAET1 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EIAET1X10 Supplier Invitrogen™ Supplier No. EIAET1X10
Add to Cart
Add to Cart
Request a Quote

ELISA

The Endothelin-1 (ET-1) Multispecies ELISA quantitates Endothelin-1 in human, bovine, procine, dog, rat, and mouse serum, plasma, urine, or cell culture medium. The assay will exclusively recognize both natural and recombinant Endothelin-1. Principle of the method The Endothelin-1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is inversely proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The protein encoded by this gene is proteolytically processed to release a secreted peptide termed endothelin 1. This peptide is a potent vasoconstrictor and is produced by vascular endothelial cells. Endothelin 1 also can affect the central nervous system. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05305
Assay Range 0.781 to 100 pg/mL
Assay Sensitivity 0.579 pg/mL
Description Endothelin-1 (ET-1) ELISA Kit
Assay Multiplex Assay on the Luminex Platform
Conjugate HRP
Detection Method Fluorescence
Product Type ELISA
Sample Type Plasma, Serum, Supernatant, Urine
Sufficient For 10 x 96 Assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 1906
Gene Symbol EDN1
Interassay CV 3.7%
Intraassay CV 3.3
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Extraction Solution
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Research Discipline Cell Signaling
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 2.5 μL, Urine 50 μL
Storage Requirements -20°C
Target Endothelin 1
Test Time 1 hrs 20 mins
Total Assay Time 2 hrs 30 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.