Learn More
CPC Scientific H-Cys-Ser-Cys-Ser-Ser-Leu-Met-Asp-Lys-Glu-Cys-Val-Tyr-Phe-Cys-His-Leu-Asp-Ile-Ile-Trp-OH (ammonium acetate salt) 1MG
SDP
Supplier:  CPC Scientific ENDT001B
Encompass
Potent vasoconstrictor that acts on endothelia cells and on the underlying smooth muscle cells as a modulator of vascular tone. ET-1 displays selectivity for ETA receptor.ONE-LETTER SEQUENCE: CSCSSLMDKECVYFCHLDIIW (C1&C15 bridge, C3&C11 bridge)MOLECULAR FORMULA: C109H159N25O32S5MOLECULAR WEIGHT:2492STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [117399-94-7], SYNONYMS: ET-1 (human, bovine, dog, mouse, porcine, rat), Endothelin-1 (human, bovine, dog, mouse, porcine, rat)RESEARCH AREA: CardiovascularREFERENCES: A. Inoue, et al., PNAS, 80, 2863 (1989); M. Yanagisawa et al., TIPS, 10, 374 (1989); G.J. Price et al., Nucleic Acids Res., 18, 3658 (1991)
Catalog No. 50-210-9236
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.