Learn More
Invitrogen™ Endothelin 1 Monoclonal Antibody (ENDO20-2101.70)

Description
MA1-10818 detects Endothelin from human samples. MA1-10818 has been successfully used in ELISA, radioimmune assay, and immunohistochemistry applications. The MA1-10818 immunogen is Human endothelin 1.
Specifications
Specifications
| Antigen | Endothelin 1 |
| Applications | ELISA, Immunohistochemistry, Radioimmune Assays (RIA) |
| Classification | Monoclonal |
| Clone | ENDO20-2101.70 |
| Concentration | 1.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | EDN1 |
| Gene Accession No. | P05305 |
| Gene Alias | ARCND3; Big endothelin 1; Big endothelin-1; Big ET; Big ET 1; Big ET1; BigET; EDN1; endothelin 1; endothelin-1; Et; ET1; ET-1; HDLCQ7; PPET1; ppET-1; preproendothelin; preproendothelin-1; preproET; QME |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.