Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Endothelin-1 Quantikine ELISA Kit
GSA_VASDP

Catalog No. SET100 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
1 Pack
Sufficient For:
1 x 96-well plate
6 x 96-well plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
SET100 1 Pack 6 x 96-well plates
DET100 1 Kit 1 x 96-well plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SET100 Supplier R&D Systems™ Supplier No. SET100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Endothelin-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Endothelin-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies multi-species Endothelin-1 in cell culture supernates (75 ul), serum (75 ul), edta plasma (75 ul), heparin plasma (75 ul), urine (75 ul).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Assay Range 0.39 - 25pg/mL
Assay Sensitivity 0.207pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. Cross-reactivity observed with 1 or more available related molecules.
Product Line R&D Systems
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine
Specificity Synthetic and Natural Endothelin-1
Sufficient For 6 x 96-well plates
Target Species Multi-species
For Use With (Application) ELISA
Quantity 1 Pack
Sample Volume Cell Culture Supernates (75uL),Serum (75uL),EDTA Plasma (75uL),Heparin Plasma (75uL),Urine (75uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym ARCND3, EDN1, endothelin 1, endothelin-1, ET1, HDLCQ7, PPET1, Preproendothelin-1, QME
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.