Learn More
Bioss Endothelin B Receptor Monoclon
SDP
Supplier:  Bioss BSM61430R
Encompass
Endothelin B Receptor Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IP RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-67399
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.