Learn More
Invitrogen™ Endothelium Monoclonal Antibody (PAL-E)

Description
The antibody only stains human and various animal blood vessels with exception of arteries. It does not stain rat, mouse and chicken endothelium. The antibody is useful for immunohistochemistry on acetone fixed frozen sections. Not useful on cellsuspensions and Western blotting.
Specifications
Specifications
| Antigen | Endothelium |
| Applications | Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | PAL-E |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Tissue culture supernatant with 10mM sodium azide |
| Gene Alias | Endothelial cell marker; Endothelial cells; Endothelium Cell Antigen-1; Pan endothelial cells |
| Host Species | Mouse |
| Immunogen | Human melanoma tissue. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.