Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUJIFILM Wako Endotoxin Extracting Solution

Catalog No. 12333041
Encompass
Change view
Click to view available options
Quantity:
4 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-333-041 4 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-333-041 Supplier FUJIFILM Wako Supplier No. 29351601
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Solution containing human serum albumin for extraction of endotoxin from medical devices

  • Capable of extracting endotoxins that are unable to be extracted in water or saline
  • For use in endotoxin testing on equipment/devices that may come into contact with fluids containing blood or protein

Specifications

Form Lyophilized
Fill Volume 10 mL
For Use With (Equipment) Equipment that come into contact with fluids containing blood or protein
Includes 4 x 10 mL vials
Quantity 4 Vials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.