Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Endotoxin Removal Resin for Wizard MagneSil Tfx™ System

Catalog No. PRA2191 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA2191 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRA2191 Supplier Promega Supplier No. A2191
May include imposed supplier surcharges.
Add to Cart
Add to Cart

The Wizard™ MagneSil™ Tfx System provides a simple method for the rapid isolation of transfection-quality plasmid DNA in a 96-well, high-throughput format.

  • Transfection-Quality Plasmid DNA in 96-Well Format
  • Improved transfection and protein expression results
  • Automated methods available

Specifications

Content And Storage 15°C to 30°C
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.