Learn More
BioVendor, LLC EndoTrap HD 10 (10 ml settled resin) Endotoxin removal resin
SDP
Supplier:  BioVendor, LLC LET0011
Encompass
EndoTrap HD 10 (10 ml settled resin) Endotoxin removal resin. EndoTrap HD removes endotoxins (lipopolysaccharides/LPS) from aqueous solutions of samples such as proteins, antibodies, vaccines, nucleic acids, buffers and cell extracts with excellent sample recovery rate (>95%). EndoTrap HD is designed to meet the needs of pharma/biotech industry for efficient removal of LPS in biomanufacturing processes and any application requiring high LPS removal rates. EndoTrap HD fulfils all requirements for industrial applications, and can easily be integrated in early or late-stage purification steps. Manufactured by LIONEX GmbH in Germany. LPS binding capacity > 5,000,000 EU/ml. Removes >99.99% of LPS. Non-toxic bacteriophage-derived ligand, no cytotoxic effects. Re-use >10 times. Sample pH range 4-10. Suitable for high salt conditions. Suitable for TRIS, HEPES, MOPS or similar buffered samples. PBS buffer possible when enriched freshly with 0.1 mM calcium (magnesium) directly before use.
Catalog No. 50-206-9078
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.