Learn More
World Precision Instrument Endozime Enzymatic Detergent, 16.9 oz.
SDP
Supplier:  World Precision Instrument WP34521
Encompass
Multi-Tiered Enzymatic Detergent with Advanced Proteolytic Action and Rust Inhibitors
Endozime AW was designed for customers who are susceptible to allergic reactions when using cleaning chemicals containing dye. It contains a proprietary formulation of protease, amylase, lipase, carbohydrase and enzymes with A.P.A. (Advanced Proteolytic Action), which removes all soil and inhibits rust. Endozime AW increases protein enzyme activity to provide a faster more thorough penetration into hard to reach places on surgical instruments and scopes.
Catalog No. NC1510634
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.