Learn More
Thomas Scientific Uvex Seismic Sealed Eyewear Black Frame Clear Lens, Anti-scratch
SDP
Supplier:  Thomas Scientific 1219G94
Encompass
Available with Dura-streme™ dual coating technology which combines our two best lens coatings — Uvextreme anti-fog on the interior and Ultra-dura anti-scratch on the exterior — for 3x longer lens life*, as well as Uvextra™ AF anti-fog or Hardcoat anti-scratch coating options Soft and flexible nasal pads conform to a variety of facial profiles and minimize slippage Soft, high-quality face cushioning and padded temple tips offer all-day comfort Highly adjustable flame-resistant headband provides excellent fit Replaceable flame-resistant, cushion-lined subframe extends the life of the eyewear Meets ANSI Z87 High Impact standard Is certified to the requirements of the CSA Z94.3 standard Uvex Seismic Sealed Eyewear…powerful eyewear for today’s demanding work environments. The Uvex Seismic provides protection against impact, sun, wind, dust and debris coupled with durable performance and minimized fogging. Its contemporary design offers a range
Catalog No. 50-146-1098
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.