Learn More
Grainger ENERGIZER Lithium Battery, 2025, 3V
Supplier:  Grainger 45EK14
Encompass_Preferred
The Energizer 2025 Lithium Battery is a 3 volt coin cell battery with a diameter of 20mm and a height of 2.5mm. It has a lithium manganese dioxide chemistry and is rated for 1200mAh capacity. The battery has a 10 year shelf life and operates from -40°F to 140°F. It is designed for devices that require low self discharge and long service life like electronics, watches, scales, laser pointers, and more. The 2025 battery provides stable voltage, high drain/pulse capability, and superior performance in extreme temperatures compared to alkaline batteries. It has a 5 year limited warranty from Energizer.
Catalog No. NC2601350
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.