Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Energy Conversion Kit
SCI_ED

Catalog No. S27886 Shop All Eisco Products
Change view
Click to view available options
Grade:
4 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S27886 4 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27886 Supplier Eisco™ Supplier No. PH1321A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Energy Conversion Kit is a great tool to discover conversion processes of different types of energy.

  • Perfect for hands on experience in the classroom
  • 3 different devices are used to generate electricity
  • 5 different devices convert the generated electricity into 5 different forms of energy
  • Includes 3 different sources for the generation of energy: hand crank (mechanical), cell battery holder (electrical), and a solar cell (light)
  • Five circuit elements can each be inserted into the board to observe the use of the generated electrical energy: motor with fan (mechanical), buzzer (sound), LED (light), resistor (heat), and an electromagnet (magnetism)
  • Apparatus measures 10 x 8 x 6.75 inches
  • Requires 2 C batteries (not included)

Specifications

Product Type Energy Conversion Kit
Grade 4 to 14
Size 10 x 8 x 6.75 in.
For Use With (Application) Discovering Conversion Processes of Different Types of Energy
Includes Hand Crank, Cell Battery Holder, Solar Cell, Motor with Fan, Buzzer, LED, Resistor, and Electromagnet
Required Accessories (2) C Batteries
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.