Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Vernier Energy Sensor
SDPSCI_ED

Catalog No. S16277ND Shop All Vernier Software Products
Change view
Click to view available options
Grade:
High School and College
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S16277ND High School and College
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16277ND Supplier Vernier™ Supplier No. VESBTA
Add to Cart
Add to Cart

Requires

Requires an interface and software, sold separately

The Vernier Energy Sensor offers an easy way to quantify voltage, current, power, and energy output of small wind turbines and solar panels such as those used in our KidWind Experiment Kits.

Source Input Potential Range: ± 30 V Source Input Current Range: ± 1000 mA

Warranty and Services

5-year limited warranty

Specifications

Description ±30 V source input potential range, ±1000 mA source input current range
Grade High School and College
Interface Required
Warranty 5 Year Limited
For Use With (Application) Educational use only
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.