Learn More
United Scientific™ Energy Transformation Balls
SDPSCI_ED
Catalog No. S000888 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Product Type:
Energy Balls
Catalog No. Product Type
S000888 Energy Balls
1 options
Prop 65
Catalog No. S000888 Supplier United Scientific™ Supplier No. ENRGYBALLS
Add to Cart
Add to Cart
Prop 65
Energy Transformation Balls transform kinetic energy into work which creates thermal energy.
  • This pair of steel balls demonstrates energy transformation.
  • By striking the two balls together with a piece of paper between them, kinetic energy is transformed into work which creates thermal energy.
  • This thermal energy creates a small burn hole in the paper.
  • There is no flame but there will be a smell of burning paper.
  • Each ball weighs 224 g (0.49 lb.) and has a diameter of 40 mm (1.5 in.).
  • Includes activity guide
  • Easily demonstrate energy transformation
  • Safely burns a hole with no flame
Specifications
Product Type Energy Balls