Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ AGP 40 cal Arc Flash Bib Overalls
SDP

Catalog No. 17010853 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Large
Large (Long)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-853 Large (Long)
17-010-852 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-010-853 Supplier National Safety Apparel™ Supplier No. ARC40BLGT
Add to Cart
Add to Cart

CAT 4 arc flash bib overalls feature a 26 in. elastic waist, leg zippers, pleated knees, and flex fit straps.

  • Single layer 40 cal arc flash protection
  • Enhanced Visibility with 3M™ Scotchlite™ FR silver reflective trim
  • Quick disconnect and adjustable Flex Fit shoulder straps
  • 26 in. leg zippers for easy donning and doffing
  • Ergonomic design for ease of movement and flexibility
  • Up to 20% lighter, 48% thinner, and 56% more flexible than other premium arc flash suits
  • Using PYRAD™ Fabric by GORE-TEX LABS comfort science to keep the wearer cool, dry, comfortable, and unrestricted while working

Specifications

Closure Type Zipper
Color Navy
Seam Type Stitched
Size Large (Long)
Suspender Adjustable
Arc Flash Rating 40 cal/cm²
Coating PYRAD™ Fabric by GORE-TEX LABS
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Product Type Bib Overalls
Certifications/Compliance NFPA 70E, CSA Z462, OSHA 1910.269, ASTM F1506
Disposable Reusable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.