Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ AGP 40 Cal Arc Flash Kit with Lift Front Hood with Fans, No Gloves
SDP

Catalog No. 17010855 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
2X-Large
3X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-855 2X-Large
17-010-856 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-010-855 Supplier National Safety Apparel™ Supplier No. ARC40KITFANNG2X
Add to Cart
Add to Cart
Request a Quote

AGP 40 cal Arc Flash Kit includes jacket, bib, lift front hood with fans and backpack.

  • Up to 20% lighter, 48% thinner, and 56% more flexible than other premium arc flash suits
  • Single layer 40 cal arc flash protection
  • Using GORE™ PYRAD™ comfort science to keep the wearer cool, dry, comfortable and unrestricted while working
  • Ergonomic design for ease of movement and flexibility
  • Covered zip front closure with large pull tab
  • 26 in. leg zippers for easy donning and doffing
  • PureView™ faceshield technology features a greater range of visibility

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Closure Type Zipper, Snap
Seam Type Stitched
Arc Flash Rating 40 cal/cm²
Suspender None
Certifications/Compliance NFPA 70E, CSA Z462, OSHA 1910.269, ASTM F1506
Color Navy
Product Type Arc Flash Kit
Size 2X-Large
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.