Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ AGP 40 Cal Arc Flash Lift Front Hood with Dual Fans
SDP

Catalog No. 17010854 Shop All National Public Seating Corporation Products
Catalog No. 17-010-854 Supplier National Safety Apparel™ Supplier No. ARC40H1
Add to Cart
Add to Cart

AGP 40 cal arc flash lift front hood has a PureView™ faceshield with anti-fog and scratch resistant coating, dual fans and a one-size hard hat.

  • Lift front shield makes it easy to obtain fresh air while out of the hazard zone
  • PureView™ faceshield technology features a greater range of visibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • Dual fans provide a gentle breeze that keeps you cool and decreases lens fogging
  • Flaps to cover fan openings when fans are removed
  • Ergonomic design for ease of movement and flexibility
  • Hard hat included
  • Up to 20% lighter, 48% thinner, and 56% more flexible than other premium arc flash suits

Specifications

Product Type Arc Flash Hood
Size One Size
Material GORE Pyrad
Color Navy
For Use With (Equipment) Utilities, Manufacturing, Electrical Contractors
Certifications/Compliance NFPA 70E, CSA Z462, OSHA 1910.269, ASTM F2178
Includes Arc Flash Hood
Style Beekeeper Hood
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.