Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI8A1, lyophilized, TrueMAB™
SDP

Catalog No. 501664208
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-4208 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-4208 Supplier OriGene Supplier No. CF506170
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

CD105 (Endoglin is a Type I transmembrane protein) is highly expressed on human vascular endothelial cells and exists on an O- and N-glycosylated homodimer. Up regulation of CD105 expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 is highly expressed on endothelial cells and promotes angiogenesis during wound healing, infarcts and in a wide range of tumors and its gene expression is stimulated by hypoxia. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2. Mutations in the CD105 gene cause hereditary hemorrhagic telangiectasia, also known as Osler-Rendu-Weber syndrome 1, an autosomal domit multisystemic vascular dysplasia. Alternatively spliced transcript variants encoding different isoforms of CD105 have been found. Diseases associated with CD105 dysfunction include hereditary hemorrhagic telangiectasia.

Specifications

Antigen ENG
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone OTI8A1
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene ENG
Gene Accession No. P17813, Q63961
Gene Alias END, HHT1, ORW1
Gene Symbols ENG
Host Species Mouse
Immunogen Full length human recombit protein of human ENG produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13805, 2022, 497010, 609166
Target Species Canine, Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.