Learn More
ENG Scientific Acid-Fast Stain Kit, Modification of the Kinyoun Method
Description
- Modification of the Kinyoun Method
- Acid-fast bacilli stain red; all other bacteria stain blue with Sol. III, green with Sol. IIIA.
- Acid Fast Stain Kit
- For in vitro diagnostic use
- 1 Kit
Specifications
Specifications
| Quantity | 1 Ea. |
| Includes | 250 mL carbol fuchsin, 250 mL acid alcohol and 250 mL Brilliant Green |
| For Use With (Application) | In vitro diagnostic use |
| Color | Dark Red |
| Stain Type | Brilliant Green |
| Form | Liquid |
| Product Type | Acid Fast Stain |
| Packaging | Kit |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.