Learn More
ENG Scientific Systain Buffer Solution
Description
- In the event of cold weather, allow solutions to return to room temperature and shake well before use.
- Contents: Wright Stain, Giemsa Stain, Methanol, Buffers, Stabilizers
- These solutions are made from certified dyes (when applicable).
Specifications
Specifications
| Quantity | 4 x 3800 mL (1 gal.) |
| Storage Requirements | Room Temperature |
| For Use With (Application) | For in-vitro diagnostic use only |
| For Use With (Equipment) | For Sysmex hematology slide stainer |
| pH | 7.2 |
| Packaging | Case (Bottles) |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.