Learn More
ENG Scientific CSF Diluting Fluid
Description
- Includes Glacial Acetic Acid, Crystal Violet
- This solution is made from certified dyes.
- For in vitro diagnostic use only.
Specifications
Specifications
| Quantity | 2 x 100 mL |
| For Use With (Application) | Hemolyzes RBCs and stains WBCs |
| Packaging | Bottles |
| Product Type | CSF Diluting Fluid |
For in vitro diagnostic use only.