Learn More
ENG Scientific Dip Stain Kit (Wright and Wright Giemsa) Components
Description
- Solutions made from certified dyes
- Solutions should be replenished during staining period
- During cold weather, allow solutions to return to room temperature and shake well before use
Specifications
Specifications
| Quantity | 3800 mL (1 gal.) |
| Storage Requirements | Room Temperature |
| For Use With (Application) | For in-vitro diagnostic use only |
| Product Type | Wright/Wright Giemsa Dip Stain - Solution I |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.