Learn More
ENG Scientific Eosin-Phloxine Solution
Catalog No. ES8922
Change view
Click to view available options
Quantity:
1 x 19 L (5 gal.)
2 x 950 mL
3.8 L (1 gal.)
950 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES8922 2 x 950 mL
ES8923 3.8 L (1 gal.)
ES8921 950 mL
ES8924 1 x 19 L (5 gal.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. ES8922 Supplier ENG Scientific Supplier No. 8922
Add to Cart
Add to Cart
This item is not returnable. View return policy
Depicts the cytoplasmic detail of the cell and the tissue structure.
  • Solution is used mainly as background contrast to nuclear stains, i.e., hematoxylin-eosin sequence
  • Made from certified dyes
Specifications
For Use With (Application) Solution is used mainly as background contrast to nuclear stains, i.e., hematoxylin eosin sequence
Quantity 2 x 950 mL
Product Type Eosin Phloxine Stain
For in vitro diagnostic use only.