Learn More
ENG Scientific Eosin-Phloxine Solution
Description
- Solution is used mainly as background contrast to nuclear stains, i.e., hematoxylin-eosin sequence
- Made from certified dyes
Specifications
Specifications
| For Use With (Application) | Solution is used mainly as background contrast to nuclear stains, i.e., hematoxylin eosin sequence |
| Packaging | Cubetainer |
| Quantity | 1 x 19 L (5 gal.) |
| Product Type | Eosin Phloxine Stain |
For in vitro diagnostic use only.