Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Gill-Hematoxylin Stain Kit (Modified)

Catalog No. ES8401
Change view
Click to view available options
Concentration:
0.2%
0.4%
0.6%
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
ES8401 0.4%
ES8400 0.2%
ES8402 0.6%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ES8401 Supplier ENG Scientific Supplier No. 8401
Add to Cart
Add to Cart

Gill-Hematoxylin Stain is a nuclear stain employing half-oxidized hematoxylin solution.

  • The stain is available in 3 concentrations at 0.2% w/v, (Modified), 0.4% w/v, and 0.6% w/v
  • The modified solution is preferred in cytology and histology
  • Gill-Hematoxylin Stain Kit (Modified)
  • Includes Ethylene Glycol, Hematoxylin, Sodium Iodate, Aluminum Sulfate, Citric Acid, Magnesium Sulfate, Sodium Bicarbonate, Ethanol
  • In vitro diagnostic use only

Specifications

Content And Storage Ethylene Glycol, Hematoxylin, Sodium Iodate, Aluminum Sulfate, Citric Acid, Magnesium Sulfate, Sodium Bicarbonate, Ethanol
Packaging Type Bottles
Concentration 0.4%
Product Type Hematoxylin
Quantity 3 x 950 mL
For Use With (Application) In vitro diagnostic use only
Test Time 2 min.

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.