Learn More
ENG Scientific Gram Stain Kit with Carbol Fuchsin Counterstain
Description
- Contains crystal violet, iodine, potassium oxalate, ethanol, acetone, safranin, basic fuchsin, phenol
- Gram-positive bacteria, many fungi and fibrin stain blue-black to violet
- Gram-negative bacteria stain red
- Cytoplasm stains pink
Specifications
Specifications
| Quantity | 4 x 250 mL |
| Includes | 250 mL crystal violet, 250 mL iodine, 250 mL potassium oxalate, 250 mL ethanol, 250 mL acetone, 250 mL safranin, 250 mL basic fuchsin, 250 mL phenol, 250 mL carbol fuchsin |
| For Use With (Application) | In vitro diagnostic use |
| Stain Type | Carbolfuchsin |
| Product Type | Gram Stain Kit |
| Packaging | Complete Kit (Bottles) |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.