Learn More
ENG Scientific Gram Stain Kit with Safranin Counterstain
Description
- Contains crystal violet, iodine, potassium oxalate, ethanol, acetone, safranin, basic fuchsin, phenol
- Gram-positive bacteria, many fungi and fibrin stain blue-black to violet
- Gram-negative bacteria stain red
- Cytoplasm stains pink
Specifications
Specifications
| Quantity | 1 x 250 mL |
| For Use With (Application) | Bacteria in tissue |
| Stain Type | Gram Stain |
| Product Type | Gram Iodine |
| Packaging | Bottle |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.