Learn More
ENG Scientific H-Pack™ May-Grunwald Giemsa Stain Packs
Description
- For AMES HEMA-TEK™ slide stainers
- To obtain desired color and intensity
- To demonstrate bacteria within leukocytes, normal and abnormal white blood cells and platelets clearly
- Freezing or high temperature does not affect staining ability
- Designed to be used at room temperature
- Offered with matched buffer solution of specific pH
- To stain LE preps, parasitic elements like malaria, thicker buffy coat smears, mature and immature elements of routine bone marrow smears, inclusion bodies like basophilic stippling
Specifications
Specifications
| Quantity | 1 Pk. |
| Storage Requirements | Room Temperature |
| For Use With (Application) | Designed to eliminate problems with precipitate, artifacts, drying of slides and overstaining |
| For Use With (Equipment) | For Ames Hema-Tek™ II slide stainers |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.