Learn More
ENG Scientific Harris' Alum Hematoxylin Stain (Mercury Free with Glacial Acetic Acid)
Description
- Staining time: 4 to 8 minutes depending on the nature of tissues
- Contents: Hematoxylin, Ethanol, Potassium Alum Hematoxylin, Ethanol, Potassium Alum, Glacial Acetic Acid
Specifications
Specifications
| Content And Storage | Hematoxylin, Ethanol, Potassium Alum Hematoxylin, Ethanol, Potassium Alum, Glacial Acetic Acid |
| Packaging Type | Bottles |
| Product Type | Hematoxylin |
| Quantity | 2 x 950 mL |
| For Use With (Application) | In vitro diagnostic use only |
| Test Time | 4 to 8 min. |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.