Learn More
ENG Scientific Heinz Body Stain
Description
Use this helpful Heinz Body Stain to recognize hemolytic anemia due to certain drugs and in detecting red cells of the primaquine-sensitive type by detecting Heinz bodies.
- Heinz bodies are best stained supravitally
- Includes Methyl Violet, Saline (NaCl)
- Made from certified dyes
Specifications
Specifications
| Quantity | 2 x 100 mL |
| For Use With (Application) | Hematology Staining |
| Packaging | Bottles |
| Product Type | Heinz Body Stain |
For in vitro diagnostic use only.