Learn More
ENG Scientific Jenner-Giema Stain Kit (May-Grunwald Giemsa Method)
Catalog No. ES8153
Change view
Click to view available options
Quantity:
1 x 100 mL
2 x 100 mL
Product Type:
Giemsa Stain
Jenner Stain
Jenner-Giemsa Stain Kit
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
ES8153 1 x 100 mL Giemsa Stain
ES8150 2 x 100 mL Jenner-Giemsa Stain Kit
ES8152 2 x 100 mL Jenner Stain
ES8151 1 x 100 mL Jenner Stain
ES8154 2 x 100 mL Giemsa Stain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ES8153 Supplier ENG Scientific Supplier No. 8153
Add to Cart
Add to Cart
Designed for staining nuclei, cytoplasm and bacteria in tissue sections.
  • Jenner-Giemsa Stain Kit
  • Kit includes Jenner stain (stock) and Giemsa stain (stock)
  • Made from certified dyes
  • For in vitro diagnostic use only
Specifications
For Use With (Application) Designed for staining nuclei, cytoplasm and bacteria in tissue sections
Includes 100 mL Jenner stain (stock) and 100 mL Giemsa stain (stock), made from certified dyes
Packaging Bottle
Quantity 1 x 100 mL
Product Type Giemsa Stain
For in vitro diagnostic use only.