Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Macchiavello Stain Kit

Catalog No. ES7250
Change view
Click to view available options
Quantity:
4 x 100 mL
4 x 250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES7250 4 x 250 mL
ES7251 4 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ES7250 Supplier ENG Scientific Supplier No. 7250
Add to Cart
Add to Cart

Differential staining procedure based on the affinity of cationic (basic) dyes for rickettsiae and chlamydiae.

  • Rickettsiae and chlamydiae stain red, tissue cells stain blue
  • For Macchiavello Stain Kit
  • Includes 250 mL of Solution I - Basic Fuchsin, Solution II - 0.5% Citric Acid, Solution III - Methylene Blue, and Solution IIIA - Malachite Green

Specifications

Quantity 4 x 250 mL
Includes 250 mL each of Solutions I, II, III and IIIA
For Use With (Application) Rickettsiae and Chlamydiae
Composition Basic Fuchsin, 0.5% Citric Acid, Methylene Blue, Malachite Green
Product Type Macchiavello Stain Kit

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.