Learn More
ENG Scientific Malarial Wright Giemsa Stain Kit
Description
- All cellular elements are stained
- The use of a thin smear allows for easy visualization of inclusions, extracellular forms and the size of red cells
- Their characteristic morphologies are used for differentiation
Specifications
Specifications
| Quantity | 3 x 950 mL |
| For Use With (Application) | For easy visualization of inclusions, extracellular forms and size of RBCs, in-vitro use only |
| Includes | Solution I Fixative, Solution II Malarial Wright Giemsa Stain, Solution III Buffer |
| Packaging | Bottles |
For in vitro diagnostic use only.