Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Maldonado's Method Stain Kit

Catalog No. ES9501
Change view
Click to view available options
Quantity:
2 x 250 mL
5 x 250 mL
Reagent Type:
Azure II (0.05%)
Complete Kit
Phloxine (1%)
Phosphotungstic Acid (3%)
Weigert's Hematoxylin
Weigert's Iron Chloride
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Reagent Type
ES9501 2 x 250 mL Phloxine (1%)
ES9500 5 x 250 mL Complete Kit
ES9502 2 x 250 mL Phosphotungstic Acid (3%)
ES9504 2 x 250 mL Weigert's Hematoxylin
ES9503 2 x 250 mL Azure II (0.05%)
ES9505 2 x 250 mL Weigert's Iron Chloride
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. ES9501 Supplier ENG Scientific Supplier No. 9501
Add to Cart
Add to Cart

For pancreatic islet cells.

  • Kit includes:
    • Solution I - Phloxine (1%)
    • Solution II - Phosphotungstic Acid (3%)
    • Solution III - Azure II (0.05%)
    • Solution IVA - Weigert's Hematoxylin
    • Solution IVB - Weigert's Iron Chloride
  • Maldonado's Method Stain Kit
  • Used with Pancreatic islet cells

Specifications

Packaging Type Bottles
Product Type Staining Reagent
Quantity 2 x 250 mL
For Use With (Application) Pancreatic islet Cells
Reagent Type Phloxine (1%)

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.