Learn More
ENG Scientific Mallory's Acid Hematoxylin Stain (Phosphotungstic Acid)
Catalog No. ES8650
Change view
Click to view available options
Quantity:
1 x 250 mL
1 x 3800 mL (1 gal.)
1 x 950 mL
2 x 950 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES8650 1 x 250 mL
ES8652 2 x 950 mL
ES8651 1 x 950 mL
ES8653 1 x 3800 mL (1 gal.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ES8650 Supplier ENG Scientific Supplier No. 8650
Only null left
Add to Cart
Add to Cart
Routine stain method for the nervous system.
  • Demonstrate abnormal or diseased astrocytes
  • Useful in showing mitochondria, striations of cardiac and voluntary muscle, and as a counterstain for the Periodic Acid-Schiff (PAS) technique
  • Recommended staining time is 12 to 24 hours
  • Mallory's Acid Hematoxylin Stain
  • Made from certified dyes
  • Includes Hematoxylin, Phosphotungstic Acid, Potassium Permanganate
Specifications
Content And Storage Hematoxylin, Phosphotungstic Acid, Potassium Permanganate
Packaging Type Bottle
Product Type Hematoxylin
Quantity 1 x 250 mL
For Use With (Application) Useful in showing mitochondria, striations of cardiac and voluntary muscle, and as a counterstain for the Periodic Acid-Schiff (PAS) technique
Test Time 12 to 24 hr.
For in vitro diagnostic use only.